missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 0.95 ± 0.01 at 25°C, Ricca Chemical

Catalog No. 148832
Change view
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
148832 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 148832 Supplier Ricca Chemical Company Supplier No. 148832
Only null left

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 0.95± 0.01 at 25°C
CAS 7647-01-0, 7732-18-5
CAS Min % 97.0, 1.2
Color Colorless
CAS Max % 100.0, 1.25
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 0.95
Physical Form Liquid
Quantity 1 L
Solution Type pH Buffer
Packaging Natural Poly Bottle
Show More Show Less