missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 0.50 ± 0.01 at 25°C, Ricca Chemical

Catalog No. R14850001A
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R14850001A 1 L
148516 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R14850001A Supplier Ricca Chemical Company Supplier No. R14850001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7647-01-0

Specifications

CAS 7647-01-0
CAS Max % 98.59, 1.41
Color Colorless Liquid
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 0.5
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Natural Poly Bottle
Sterility Non-sterile
Grade Not Available
Show More Show Less