missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer, Precision Reference Standard, pH 8.000 ± 0.005 at 25°C, Ricca Chemical
Specifications
Specifications
| CAS | 7732-18-5, 7558-79-4, 7778-77-0 |
| CAS Max % | 99.05, 0.9, 0.04 |
| Color | Colorless Liquid |
| Concentration | 1X |
| For Use With (Application) | Calibration |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 8 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Solution Type | pH Buffer |
| Show More |