missing translation for 'onlineSavingsMsg'
Learn More

Buffer, pH 8.00, USP Reagent, Ricca Chemical

Catalog No. R15832001A
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R15832001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R15832001A Supplier Ricca Chemical Company Supplier No. R15832001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Buffer, pH 8.00
Name Note PH 8.00
CAS 10043-35-3, 1310-73-2, 7447-40-7, 7732-18-5
Color Colorless
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 8
Physical Form Liquid
Quantity 1 L
Solution Type pH Buffer
Odor Odorless
Packaging Natural Poly Bottle
Solubility Information Solubility in Water: Infinite
Grade USP
Show More Show Less