missing translation for 'onlineSavingsMsg'
Learn More

Buffer pH 7, Green, Reagents

Catalog No. 01335734
Change view
Click to view available options
Quantity:
500 mL
4 L
10 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335734 10 L
01335733 20 L
01335732 4 L
01335731 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01335734 Supplier Labchem Inc Supplier No. CB16750010F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Buffer pH 7, Green, Reagent, use for pH Meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Specifications

Chemical Name or Material Buffer pH 7, Green
CAS 7732-18-5, 7558-79-4, 7558-79-4
Boiling Point 100°C
Color Green
For Use With (Application) Calibration
For Use With (Equipment) pH Meter
pH 7
Physical Form Liquid
Quantity 10 L
Solution Type Buffer
Packaging Cubitainer
Specific Gravity 1.006
Sterility Non-sterile
Grade Reagent
Show More Show Less