missing translation for 'onlineSavingsMsg'
Learn More

Buffer pH 5.00 ± 0.01 at 25°C Reference Standard, Reagents

Catalog No. 01337454
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01337454 4 L Poly Bottle
01337457 10 L Cubitainer
01337456 1 L Poly Bottle
01337455 20 L Cubitainer
01337453 4 L Cubitainer
01337452 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01337454 Supplier Labchem Inc Supplier No. C21552504A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Buffer pH 5.00 ± 0.01 at 25°C Reference Standard, Reference Buffer, use for pH meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Specifications

Chemical Name or Material Buffer pH 5.00 ± 0.01 at 25°C Reference Standard
CAS 7732-18-5, 877-24-7, 1310-73-2
Melting Point 0°C
Boiling Point 100°C
Color Colorless
For Use With (Application) Calibration
For Use With (Equipment) pH Meter
pH 5
Physical Form Liquid
Quantity 4 L
Solution Type Buffer
Packaging Poly Bottle
Specific Gravity 1.007
Sterility Non-sterile
Grade Not Available
Show More Show Less