missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer concentrate pH 12.00, For 500 ml buffer solution, di-sodium hydrogen phosphate / sodium hydroxide solution, Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Chemical Name or Material | Buffer concentrate |
| pH | 12 |
| Quantity | 1 Pc. |