missing translation for 'onlineSavingsMsg'
Learn More

Buffer Concentrate, pH 10.00 (Certified), Fisher Chemical

Catalog No. SB141500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB141500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB141500 Supplier Thermo Fisher Scientific Supplier No. FLSB141500
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Buffer Concentrate, pH 10.01
CAS 7447-40-7, 1310-73-2, 139-33-3, 7732-18-5, 10043-35-3
Color Undesignated
pH 10.0
Physical Form Liquid
Quantity 500 mL
Identification Pass Test
MDL Number MFCD00163176
Packaging Poly Bottle
Traceability to NIST Traceable to NIST.
Grade Certified
Show More Show Less