missing translation for 'onlineSavingsMsg'
Learn More

Budesonide Related Compound G Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013012
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013012 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013012 Supplier Merck Emd Millipore Supplier No. PHR182250MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 137174-25-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C25H36O6
Synonym 16 alpha,17-[Butylidenebis(oxy)]-11 beta, 21-dihydroxypregna-4-ene-3,20-dione; 16 alpha,17-[butylidenebis(oxy)]-11 beta, 21-dihydroxypregna-4-ene-3,20-dione; 1,2-Dihydrobudesonide
Type Certified Reference Material
Quantity 50 mg
Formula Weight 432.55
Grade Certified Reference Material