missing translation for 'onlineSavingsMsg'
Learn More

Budesonide Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014400
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014400 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014400 Supplier Merck Emd Millipore Supplier No. PHR1178500MG
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Budesonide is a nonsystemic glucocorticoid and an epimeric mixture, where both the epimers have similar anti-inflammatory activity. It is widely used in the treatment of Crohn′s disease. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 51333-22-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C25H34O6
MDL Number MFCD00083259
Synonym 16,17-Butylidenebis(oxy)-11,21-dihydroxypregna-1,4-diene-3,20-dione
Type Certified Reference Material
Quantity 500 mg
Formula Weight 430.53
Grade Certified Reference Material