missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Polypropylene Buchner Funnels

Two piece polypropylene Buchner funnels are light weight and autoclavable. The top portion of the funnel cone is fitted with the lower portion of the funnel cone forming a vacuum seal that does not require greasing. Bottom portion of the funnel cone has a perforated filter plate.

$30.32 - $46.29

Specifications

Material Polypropylene
Autoclavable Autoclavable
Color White
Description Buchner Funnel
For Use With (Application) Filtering liquids
View More Specs

Products 3
Catalog Number Mfr. No. Diameter (Metric)Top Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric)Top Price Quantity & Availability  
S95946
View Documents Promotion Details
Eisco™
CH0464A
70 mm
Each for $37.68
Only null left
 
S95947
View Documents Promotion Details
Eisco™
CH0464B
110 mm
Each for $46.29
Only null left
 
S89263
View Documents Promotion Details
Eisco™
CH0464C
90 mm
Each for $30.32
Only null left
 
Description

Description

  • Two-piece Buchner funnel with plain stem
  • Made from polypropylene
  • Perforated filter plate on base of cone
  • No greasing necessary
  • Autoclavable

Warranty

1 year limited manufacturer warranty.

Specifications

Specifications

Polypropylene
White
Filtering liquids
1
Polypropylene
Autoclavable
Buchner Funnel
Filtration paper
Buchner Funnel
Videos
SDS
Documents

Documents