missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BUBR1 Recombinant Rabbit Monoclonal Antibody (23GB1640)
GREENER_CHOICE

Catalog No. MA554122
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA554122 100 μL
MA554123 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA554122 Supplier Invitrogen™ Supplier No. MA554122
Only null left

Rabbit Recombinant Monoclonal Antibody

BUBR1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

BUBR1 is a member of the Ser/Thr protein kinase family. It is a component of the mitotic checkpoint that delays anaphase until all chromosome are properly attached to the mitotic spindle. Compromised spindle checkpoints are thought to play an important role in genetic instability that predisposes cells to malignant transformations. Human cells contain two related checkpoint protein kinases, evolved from the BUB1 gene; hBUBR1 and hBUB1. They are important components of the spindle checkpoint Both kinases monitor mitotic kinetochore-microtubule interactions. hBUBR1 is essential for normal mitotic progression as it prevents cells from entering into anaphase too early. BUBR1 has been detected in human cancers. It has been identified as a novel binding partner of BCSG1 (Breast Cancer-Specific Gene 1), which has been suggested to accelerate the progression of breast cancer by compromising the mitotic checkpoint control through inactivation of BUBR1.

Specifications

Antigen BUBR1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1640
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BUB1B
Gene Accession No. O60566
Gene Alias AU045529; BUB1 budding uninhibited by benzimidazoles 1 homolog beta; BUB1 mitotic checkpoint serine/threonine kinase B; Bub1A; Bub1b; BUB1B, mitotic checkpoint serine/threonine kinase; BUB1beta; BUBR1; budding uninhibited by benzimidazoles 1 beta; budding uninhibited by benzimidazoles 1 homolog beta; budding uninhibited by benzimidazoles 1 homolog, beta (S. cerevisiae); hBub1; hBUBR1; MAD3/BUB1-related protein kinase; MAD3L; Mitotic checkpoint kinase MAD3L; mitotic checkpoint serine/threonine-protein kinase BUB1 beta; MVA1; Protein SSK1; SSK1
Gene Symbols BUB1B
Host Species Rabbit
Immunogen A synthesized peptide derived from human BubR1 (600-650AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 701
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less