missing translation for 'onlineSavingsMsg'
Learn More

BTX™ BTXPress™ Electroporation Buffer Kit

BTXPress™ Electroporation Buffer Kit offers a single buffer solution developed to quickly and efficiently deliver genes into mammalian cells that were previously considered hard to transfect by chemical and other non-viral methods.

$1398.96 - $2642.75

Specifications

Content And Storage 2°C to 8°C
Cell Type Mammalian
Promoter None (Promoterless)
Transfection Technique Electroporation
For Use With (Application) Transfection via Electroporation
View More Specs

Products 4
Catalog Number Mfr. No. Quantity No. of Reactions Price Quantity  
Catalog Number Mfr. No. Quantity No. of Reactions Price Quantity  
BTX450803
View Documents
BTX™
450803
5 Bags (10 Cuvettes/Bag) 50
Each for $1,909.84
Only null left
 
BTX450806
View Documents
BTX™
450806
2 Bags (50 Cuvettes/Bag) 100
Each for $2,642.75
Only null left
 
BTX450804
View Documents
BTX™
450804
2 Bags (10 Cuvettes/Bag) 20
Each for $1,398.96
Only null left
 
BTX450807
View Documents
BTX™
450807
4 Bags (10 Cuvettes/Bag), 10 mL Buffer 40
Each for $2,209.46
Only null left
 
Description

Description

BTXPress™ Electroporation Solution is the first electroporation reagent that meets all of your high performance transfection needs without sacrificing control over your experiment or your budget. It offers increased numbers of transfections per kit compared to other commercial alternatives, providing higher value to you. As a universal solution, the BTXPress™ Electroporation Solution can be used in other electroporators, including the Amaxa™, to deliver similar results without the typical high cost associated with these buffer kits. The BTXPress™ Electroporation Solution is offered as a kit including the BTX™ cuvettes Plus with transfer pipettes or as a buffer alone.

  • Efficient electroporation transfection
  • High efficiency transfection of difficult cell lines
  • High cell viability
  • Low toxicity resulting in improved cell viability
  • Single buffer for all cell types
  • One buffer used in place of standard electroporation buffers for all mammalian cell types
Specifications

Specifications

2°C to 8°C
None (Promoterless)
Transfection via Electroporation
Electroporation Buffer
Mammalian
Electroporation
BTX Electroporation
Sterile
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'