missing translation for 'onlineSavingsMsg'
Learn More

BTX™ BTXPress™ Electroporation Buffer

Catalog No. BTX450801
Change view
Click to view available options
Quantity:
2 mL
5 mL
10 mL
No. of Reactions:
100
20
50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity No. of Reactions
BTX450801 2 mL 20
BTX450802 5 mL 50
BTX450805 10 mL 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BTX450801 Supplier BTX™ Supplier No. 450801
Only null left

BTXPress™ Electroporation Buffer is a single buffer solution developed to quickly and efficiently deliver genes into mammalian cells that were previously considered hard to transfect by chemical and other non-viral methods.

BTXPress™ Electroporation Solution is the first electroporation reagent that meets all of your high performance transfection needs without sacrificing control over your experiment or your budget. It offers increased numbers of transfections per kit compared to other commercial alternatives, providing higher value to you. As a universal solution, the BTXPress™ Electroporation Solution can be used in other electroporators, including the Amaxa™, to deliver similar results without the typical high cost associated with these buffer kits. The BTXPress™ Electroporation Solution is offered as a kit including the BTX™ cuvettes Plus with transfer pipettes or as a buffer alone.

  • Efficient electroporation transfection
  • High efficiency transfection of difficult cell lines
  • High cell viability
  • Low toxicity resulting in improved cell viability
  • Single buffer for all cell types
  • One buffer used in place of standard electroporation buffers for all mammalian cell types

Specifications

Buffer Universal Electroporation Buffer
Content And Storage 2°C to 8°C
Color Clear
Description BTXpress Solution, 2.0 ml Bottle for up to 20 Reactions
Cell Type Mammalian
Format Bottle
Sample Type Cells
Promoter None (Promoterless)
Transfection Technique Electroporation
For Use With (Equipment) BTX™ and other electroporators
For Use With (Application) Transfection via Electroporation
Includes 2 mL Buffer only
Packaging Type Bottle
No. of Reactions 20
Product Line BTX Electroporation
Product Type Electroporation Buffer
Quantity 2 mL
Sterility Sterile
Volume (Metric) 2 mL
Show More Show Less