missing translation for 'onlineSavingsMsg'
Learn More

BTX™ BTXPress™ Cytoporation Low Conductivity Medium T4

Catalog No. BTX470003
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTX470003 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTX470003 Supplier BTX™ Supplier No. 470003

BTXPress™ Cytoporation Low Conductivity Medium T4 is an advanced electroporation buffer designed for use with the BTX™ AgilePulse™ MAX Large Volume Electroporation System for ex vivo or in vitro delivery of DNA, RNA, oligonucleotides, and siRNA.

  • Sterile-filtered from the highest quality non-animal, medical-grade reagents
  • Buffer can be directly diluted in complete growth media for post-electroporation cell culture
  • Optimized for eukaryotic electrofusion applications
  • Maximum transfection efficiency, high cell viability
  • Low conductivity for minimal heating
  • Physiological pH and balanced osmolarity
  • Contains no animal products
  • Certified RNase and DNase free

Specifications

Format 500 mL Bottle
For Use With (Application) Ex vivo or in vitro delivery of DNA, RNA, oligonucleotides, and siRNA
For Use With (Equipment) BTX AgilePulse MAX Large Volume Electroporation Systems
Product Line BTX Electroporation
Product Type Electroporation Consumable
Description BTXpress Cytoporation Low Conductivity Medium T4
Life Span 18 months from date of manufacture
Quantity 500 mL
Sterility Sterile
Content And Storage 2°C to 8°C
Pulse Form Square Wave
Show More Show Less