missing translation for 'onlineSavingsMsg'
Learn More

BTX™ BTXPress™ Cytoporation Low Conductivity Medium T

Catalog No. BTX470002
Change view
Click to view available options
Connection Type:
Chamber Cable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
BTX470002 Chamber Cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTX470002 Supplier BTX™ Supplier No. 470002

BTXPress™ Cytoporation Low Conductivity Medium T is an advanced electroporation buffer designed for use with the BTX™ AgilePulse™ MAX Large Volume Electroporation System for ex vivo or in vitro delivery of DNA, RNA, oligonucleotides, and siRNA.

  • Sterile-filtered from the highest quality non-animal, medical-grade reagents
  • Buffer can be directly diluted in complete growth media for post-electroporation cell culture
  • Optimized for eukaryotic electrofusion applications
  • Maximum transfection efficiency, high cell viability
  • Low conductivity for minimal heating
  • Physiological pH and balanced osmolarity
  • Contains no animal products
  • Certified RNase and DNase free

Specifications

Format Large volume electroporation
For Use With (Application) Ex vivo or in vitro delivery of DNA, RNA, oligonucleotides, and siRNA
For Use With (Equipment) BTX AgilePulse MAX Large Volume Electroporation System
Product Line BTX Electroporation
Product Type Electroporation Consumable
Description BTXpress Cytoporation Low Conductivity Medium T
Life Span 18 months from date of manufacture
Quantity 500 mL
Sterility Sterile
Content And Storage 2°C to 8°C
Connection Type Chamber Cable
Pulse Form Square Wave
Show More Show Less