missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Two-Needle Array™ Electrodes: Handle Only
Description
Includes
8cm long array handle
s
- Reusable
- 45-0167 handle designed for 45-0120 10 mm needle tips
- Used for larger muscle masses
- 45-0168 handle designed for 45-0121 5 mm needle tips
- Recommended for smaller muscle masses
Specifications
Specifications
| For Use With (Application) | In-Vivo Drug or Gene Delivery, Muscle Gene Therapy |
| For Use With (Equipment) | 2-needle Array (45-0168), ECM 830, 2001, 2001+ and Gemini X2 Generators |
| Material | Delrin |
| Product Line | BTX Electroporation |
| Product Type | Electroporation accessory |
| Voltage | 500 V |
| Description | 2-Needle Array Handle, 5 mm |
| Includes | Handle only. Requires 45-0168 |
| Pulse Length Range | 10 μsec. to 99 msec. in PBS |
| Quantity | 1 Ea. |
| Show More |