missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Two-Needle Array™ Electrodes: Handle Only

Catalog No. BTX530
Change view
Click to view available options
Size (Metric) Gap:
5 mm
10 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric) Gap
BTX530 10 mm
BTX532 5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTX530 Supplier BTX™ Supplier No. 45 0205

Includes

8cm long array handle
s

Handle for use with BTX Two-Needle Array™ Electrodes (cat. no. BTX531).

  • Reusable
  • 45-0167 handle designed for 45-0120 10 mm needle tips
    • Used for larger muscle masses
  • 45-0168 handle designed for 45-0121 5 mm needle tips
    • Recommended for smaller muscle masses

Specifications

For Use With (Application) In-Vivo Drug or Gene Delivery, Muscle Gene Therapy
For Use With (Equipment) 2-needle Array (45-0167), ECM830, Gemini X2 and ECM2001+ Generators
Material Delrin
Product Line BTX Electroporation
Product Type Electroporation accessory
Voltage 500 V
Description 2-Needle Array Handle, 10 mm
Includes Electrode handle only (requires 45-0167)
Length (English) 3.2 in.
Length (Metric) 80 mm
Pulse Length Range 10 μsec. to 99 msec. in PBS
Quantity 1 Ea.
Size (English) Gap 0.39 in.
Size (Metric) Gap 10 mm
Sterility Non-sterile
Connection Type Banana Cables
Pulse Form Square Wave
Required Accessories 2-needle Array (45-0167)
Show More Show Less