missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Two-Needle Array™ Electrodes

Catalog No. BTX533
Change view
Click to view available options
Size (Metric) Gap:
5 mm
10 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric) Gap
BTX533 5 mm
BTX531 10 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTX533 Supplier BTX™ Supplier No. 45 0121

Two-Needle Array™ Electrodes designed for in vivo drug or gene delivery.

  • Sterile, disposable two-needle array
  • Enables simple, inexpensive, safe direct injection of muscle tissue for plasmid DNA delivery
  • Compatible with ECM™ 830, ECM 2001, ECM 630

Two Sizes

  • 5 mm array and handle recommended for small muscle masses such as mouse tibialis
  • 10 mm size suitable for larger muscle masses such as rat gastronomes

Specifications

For Use With (Application) In-Vivo Drug or Gene Delivery, Muscle Gene Therapy
For Use With (Equipment) ECM 830, ECM 2001+, ECM 630, Legacy ECM 2001 Generator
Material Medical Grade Stainless Steel
Product Line BTX Electroporation
Product Type Electroporation Accessory
Voltage 500 V
Description 2-Needle Array, 5 mm Gap, Pkg. of 6
Electrode Shape Straight/Needle
Includes Pk. 6 Needle Array. Handle and connectors required
Length (English) Electrode 0.78 in.
Length (Metric) Electrode 20 mm
Pulse Length Range 10 μsec. to 99 msec. in PBS
Quantity 6 Pk.
Size (English) Gap 0.19 in.
Size (Metric) Gap 5 mm
Sterility Sterile
Connection Type Banana Plug to Banana Plug
Pulse Form Square Wave
Required Accessories Handle, Connectors
Show More Show Less

Intended for Research use only. Not for use in diagnostic or clinical procedures.