Learn More
BTX™ Safety Stand for Electroporation
Safety Stand compatible with BTX™ Gemini X2, ECM 399, ECM 630 and ECM 830 Electroporation Systems.
Supplier: BTX™ 450207
Description
- For use with disposable Fisher cuvettes
- Unique design ensures no current unless the lid is closed
- Up to two cuvettes can be electroporated simultaneously
- Separate designs for Gemini X2 and Gemini SC
Specifications
| Disposable Cuvettes, Holds Various Types of Chambers/Two Cuvettes at The Same Time | |
| BTX Electroporation | |
| Safety Stand, Adjustable Gap (for use with Cuvettes) | |
| 1 Ea. | |
| Banana Cables |
| Gemini X2, ECM 399, ECM 630, ECM 830 | |
| Safety Stand | |
| 4 mm Banana Cables | |
| Non-sterile | |
| Exponential Decay Wave, Square Wave |