missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Genetrodes™ Electrode Kit

Genetrodes™ Electrode Kit features needle-style reusable electrodes. Available in five models to suit your application needs.

Supplier:  BTX™ 45 0161

 View more versions of this product

Catalog No. BTX450161


Only null left

Includes: 45-0203 Genetrodes/Genepaddle Holder, 45-0201 Square Post Cables and 45-0210 Coaxial Banana Plug Cable

Description

Description

  • Needle-style reusable electrodes compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
  • Five models available
  • Models 508 and 510 are straight with gold tips designed for general in vivo applications
  • Models 512, 514, and 516 are L-shaped electrodes with gold tips designed for in ovo electroporation
  • 515 Genetrodes Holder correctly positions all electrodes at a predetermined gap (10 grooves, 1 mm apart)
Specifications

Chemical Identifiers

ECM 830, ECM 2001+, ECM 2001, and Gemini X2
200 V
0.5 mm
1 sec. to 99 msec.
1 Ea.
1 to 10 mm
Non-sterile
Square Wave
Gold
0.02 in.
∼1500+ Pulses
10 μsec. to 99 msec. in PBS
0.03 to 0.39 in.
Needle Style
Banana Plug to Micrograbber

Specifications

In vivo, ex vivo, in ovo gene delivery applications
Gold
Genetrodes Kit, 10 mm Straight (GOLD TIP)
0.5 mm
Genetrodes/Genepaddle Holder (45-0203), Square Post Cables (45-0201), Coaxial Banana Plug Cable (45-0210)
10 mm
1 sec. to 99 msec.
1 Ea.
1 to 10 mm
Non-sterile
Square Wave
ECM 830, ECM 2001+, ECM 2001, and Gemini X2
200 V
0.02 in.
Straight
0.39 in.
∼1500+ Pulses
10 μsec. to 99 msec. in PBS
0.03 to 0.39 in.
Needle Style
Banana Plug to Micrograbber
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Intended for Research use only. Not for use in diagnostic or clinical procedures.