missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Caliper Electrodes

Catalog No. BTX384L
Change view
Click to view available options
Length (Metric):
1 cm
2 cm
Width (Metric):
1 cm
2 cm
Material:
Brass
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Width (Metric) Material
BTX384L 2 cm 2 cm Stainless Steel
BTX384 1 cm 1 cm Brass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTX384L Supplier BTX™ Supplier No. 45 0102

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Caliper Electrodes for in-vivo electroporation.

  • For in vivo electroporation, specifically the delivery of molecules or drugs into a desired tissue
  • Consist of two stainless-steel electrodes that grasp the surface site to be electroporated
  • Applications include:
    • Electrochemotherapy (ECT): the use of pulsed electric fields to deliver chemotherapeutic drugs into cancerous tumors or
    • Transdermal Delivery (TDD): use of a pulsed electric field to deliver drugs or other molecules through the skin to the targeted site
  • For use with BTX Gemini X2 electroporation systems.

Specifications

For Use With (Application) In-Vivo Drug or Gene Delivery, Electrochemotherapy, Transdermal Applications, Intact Plant Applications
For Use With (Equipment) ECM 830, Gemini X2, ECM 2001+, and Legacy ECM 2001
Material Stainless Steel
Product Line BTX Electroporators
Product Type Electroporation accessory
Voltage 500 V
Description Caliper Electrode, 2.0 cm x 2.0 cm with extra 1.5 cm x 1.5 cm electrodes included, Stainless Steel
Electrode Shape Square
Includes Caliper Electrode, 2.0 cm x 2.0 cm with extra 1.5 cm x 1.5 cm electrode included
Length (English) 0.79 in.
Length (Metric) 2 cm
Life Span Unlimited when properly maintained
Pulse Length Range 10 μsec. to 99 msec.
Quantity 2 Pk.
Size (English) Gap 0.039 to 5.11 in.
Size (Metric) Gap 1 to 130 mm
Tip Type Stainless Steel Caliper
Width (English) 0.79 in.
Width (Metric) 2 cm
Sterility Non-sterile
Connection Type Banana Cables
Pulse Form Square Wave
Show More Show Less

Intended for Research use only. Not for use in diagnostic or clinical procedures.