missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Tweezertrode™ Electrode Kit

Catalog No. BTX450165
Change view
Click to view available options
Diameter (Metric):
7 mm
10 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
BTX450165 7 mm
BTX450166 10 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTX450165 Supplier BTX™ Supplier No. 45 0165

Tweezertrode™ Electrode Kit features reusable, tweezer-style, in vivo electrodes suitable for drug or gene delivery in animals.

  • Consist of a standard 11.5 cm tweezer modified with disk electrodes of stainless steel or platinum at the tip
  • Compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
  • Gap between electrode disks may be adjusted from 1 mm to more than 2 cm
  • Can be easily cleaned
  • Kits are supplied with BTX524 coaxial cable for connection to electroporation generators

Specifications

For Use With (Application) In vivo applications, including in utero and ex-vivo gene transfection, eletroporation therapy, and transdermal drug delivery.
For Use With (Equipment) ECM 830, Gemini X2, ECM 2001+, Legacy ECM 2001 Generator
Material Stainless Steel
Product Line BTX Electroporation
Product Type Electrode Kit
Voltage 1000 V
Description Stainless Tweezertrode Kit, 7 mm
Diameter (English) 0.27 in.
Diameter (Metric) 7 mm
Electrode Shape Circular
Includes Cable
Length (English) 0.27 in.
Length (English) Electrode 0.47 in.
Length (Metric) 7 mm
Length (Metric) Electrode 12 mm
Pulse Length Range 10 μsec. to 99 msec. in PBS
Quantity 1 Ea.
Relative Humidity 20 to 80%
Size (English) Gap <0.04 in. to >0.79 in.
Size (Metric) Gap <1 mm to >2 cm
Tweezer Size 11.5 cm
Sterility Non-sterile
Connection Type Banana Cables
Pulse Form Square Wave
Show More Show Less

Intended for Research use only. Not for use in diagnostic or clinical procedures.