missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Tweezertrode™ Electrode
Tweezertrode™ Electrode is a reusable, tweezer-style, in vivo electrode suitable for drug or gene delivery in animals.
Supplier: BTX™ 450486
Description
- Consist of a standard 11.5 cm tweezer modified with disk electrodes of stainless steel
- Compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
- Gap between electrode disks may be adjusted from 1 mm to more than 2 cm
- Can be easily cleaned
Order Info
Tweerzertrode kits (Cat. No. BTX450486) and additional cable (BTX524) also available separately.
Specifications
| Vivo applications, including in utero and ex-vivo gene transfection, eletroporation therapy, and transdermal drug delivery. | |
| Platinum | |
| Tweezer Style Electrode | |
| Platinum Tweezertrode, 1 mm Diameter | |
| 1 mm | |
| 0.47 in. | |
| 41°F to 104°F | |
| 1 μsec. to 200 msec. | |
| 1 Ea. | |
| Adjustable: < 1 mm to > 2 cm | |
| 11.5 cm | |
| Banana Cables |
| BTX 830, ECM 2001+, Gemini X2 Generators | |
| BTX Electroporation | |
| 200 V | |
| 0.039 in. | |
| Circular | |
| 11.9 mm | |
| 1 sec. to 99 msec. | |
| 5°C to 40°C | |
| 20 to 80% | |
| Tweezer Style | |
| Non-sterile | |
| Square Wave |
Intended for Research use only. Not for use in diagnostic or clinical procedures.