missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Tweezertrode™ Electrode

Catalog No. BTX522
Change view
Click to view available options
Diameter (Metric):
1 mm
3 mm
5 mm
7 mm
10 mm
Material:
Platinum
Stainless Steel
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Material
BTX522 10 mm Stainless Steel
BTX450486 1 mm Platinum
BTX450487 3 mm Platinum
BTX450489 5 mm Platinum
BTX450488 7 mm Platinum
BTX520 7 mm Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. BTX522 Supplier BTX™ Supplier No. 45 0119

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tweezertrode™ Electrode is a reusable, tweezer-style, in vivo electrode suitable for drug or gene delivery in animals.

  • Consist of a standard 11.5 cm tweezer modified with disk electrodes of stainless steel
  • Compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
  • Gap between electrode disks may be adjusted from 1 mm to more than 2 cm
  • Can be easily cleaned

Order Info

Tweerzertrode kits (Cat. No. BTX450486) and additional cable (BTX524) also available separately.

Specifications

For Use With (Application) In vivo applications, including in utero and ex-vivo gene transfection, eletroporation therapy, and transdermal drug delivery.
For Use With (Equipment) ECM 830, ECM 2001+, Legacy ECM2001, Gemini X2
Material Stainless Steel
Product Line BTX Electroporation
Product Type Tweezer Style Electrode
Voltage 1000 V
Description Stainless Tweezertrode Electrode, 10 mm Diameter
Diameter (English) 0.393 in.
Diameter (Metric) 10 mm
Electrode Shape Circular
Pulse Length Range 10 μsec. to 99 msec. in PBS
Quantity 1 Ea.
Tip Type Tweezer Style
Tweezer Size 11.5 cm
Sterility Non-sterile
Connection Type Banana to Pin Tip
Pulse Form Square Wave
Required Accessories Adaptor Cable
Show More Show Less

Intended for Research use only. Not for use in diagnostic or clinical procedures.