missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Tweezertrode™ Electrode

Tweezertrode™ Electrode is a reusable, tweezer-style, in vivo electrode suitable for drug or gene delivery in animals.

Supplier:  BTX™ 45 0118

 View more versions of this product

Catalog No. BTX520


Only null left

Description

Description

  • Consist of a standard 11.5 cm tweezer modified with disk electrodes of stainless steel
  • Compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
  • Gap between electrode disks may be adjusted from 1 mm to more than 2 cm
  • Can be easily cleaned

Order Info

Tweerzertrode kits (Cat. No. BTX450486) and additional cable (BTX524) also available separately.

Specifications

Chemical Identifiers

BTX Electroporation
Circular
Tweezer Style
Non-sterile
Tweezer Style Electrode
1 Ea.
11.5 cm
Square Wave

Specifications

In vivo applications, including in utero and ex-vivo gene transfection, eletroporation therapy, and transdermal drug delivery.
Stainless Steel
Tweezer Style Electrode
Stainless Tweezertrode Electrode, 7 mm Diameter
7 mm
10 μsec. to 99 msec. in PBS
Tweezer Style
Non-sterile
Square Wave
ECM 830, ECM 2001+, Gemini X2, and Legacy ECM 2001
BTX Electroporation
1000 V
0.27 in.
Circular
1 Ea.
11.5 cm
Banana to Pin Tip
Adaptor Cable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Intended for Research use only. Not for use in diagnostic or clinical procedures.