missing translation for 'onlineSavingsMsg'
Learn More

BTX™ HT 25-well Electroporation Plates

Catalog No. BTXHTP252
Change view
Click to view available options
Size (Metric) Gap:
2 mm
Volume (Metric):
100 μL (Working)
Quantity:
1 Ea.
6 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric) Gap Volume (Metric) Quantity
BTXHTP252 2 mm 100 μL (Working) 1 Ea.
BTX450467 2 mm 100 μL (Working) 6 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTXHTP252 Supplier BTX™ Supplier No. 45 0466

HT 25-well Electroporation Plates for use with ECM 630 and ECM 830 Electroporation Systems.

  • 25-well disposable electroporation plate
  • Standard multi-well formats
  • Bio-compatible materials

Specifications

Format 25-well
For Use With (Application) siRNA Transfection, Library Studies and Stem Cell Project
For Use With (Equipment) ECM 630HT, ECM 830HT, Gemini X2 HT Electroporation System
Material Medical grade polycarbonate, polished aluminium
Product Line BTX Electroporation
Product Type Electroporation consumables
Description 25-Well Disposable Electroporation Plate, 2 mm gap, 100 μl, coated, 1 plate
Electroporation Chamber 25-well Electroporation Plate
Includes 1 x plate with seal
Quantity 1 Ea.
Size (English) Gap 0.078 in.
Size (Metric) Gap 2 mm
Volume (Metric) 100 μL (Working)
Sterility Sterile
Pulse Form Exponential Decay Wave, Square Wave
High-throughput Compatibility Yes
Show More Show Less