missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Legacy ECM 830 Foot Switch (compatible with non-touchscreen models)

Catalog No. BTX1250FS
Change view
Click to view available options
Connection Type:
Output Cable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
BTX1250FS Output Cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTX1250FS Supplier BTX™ Supplier No. 45 0211

Legacy ECM 830 Foot Switch allows hands-free operation.

  • Functions in place of the start button on the front of the generator
  • Ideal when conducting in vivo/in ovo gene delivery or nuclear transfer experiments when both hands are needed for sample manipulation
  • Compatible with BTX generators as specified in table1

1Also compatible with some older model units. Contact your Fisher Scientific™ Customer Service Representative for more information.

Specifications

For Use With (Application) In-Vivo Gene Delivery, in-Ovo Gene Delivery and Nuclear Transfer/Cloning
For Use With (Equipment) Legacy ECM 830 Square Wave Electroporation System (Non-touch screen models)
Product Line BTX Electroporation
Product Type Electroporation accessory
Description Legacy ECM 830 Foot Pedal (compatible with non-touchscreen models)
Depth (English) 3.5 in.
Depth (Metric) 89 mm
Height (English) 2 in.
Height (Metric) 51 mm
Length (English) Cable 10 ft.
Length (Metric) Cable 3.1 m
Width (English) 4.5 in.
Width (Metric) 114 mm
Sterility Non-sterile
Connection Type Output Cable
Interface Footswitch
Pulse Form Square Wave
Show More Show Less