missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Banana to Banana Cable, Red and Black, 10 ft.

Connect external devices (Genetrodes, Genepaddles, Microslides, Meander Fusion Chamber, Flat electrode) to BTX electroporation generators with this Banana to Banana Cable.

Supplier:  BTX™ 450217

Catalog No. BTX450217

Only null left
Explore available promotions

Description

Description

  • Compatible with most BTX Systems such as ECM 2001+ and Gemini
  • See the electrode type for more details
Specifications

Specifications

Required to Connect The Flat Electrode, 2 needle array and Petri pulser to The Electroporation Generator
Flat Electrode Connector Cable
10 ft.
Non-sterile
AC Sine Wave, DC Square Wave
ECM 830, Gemini X2, ECM 2001 Generator
Banana to Banana Cables, Red and Black, 10 ft
3.1 m
Banana Plug to Banana Plug
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions