missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BTNL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113828
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113828 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113828 Supplier Invitrogen™ Supplier No. PA5113828
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: EDWESKQMPQ YRGRTEFVKD SIAGGRVSLR LKNITPSDIG LYGCWFSSQI For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 77%; Horse: 77%; Human: 100%; Pig: 77%.

BTNL3 is expressed in small intestine, colon, testis, spleen, and leukocyte and plays a role in lipid metabolism. Two isoforms have been described.

Specifications

Antigen BTNL3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene BTNL3
Gene Accession No. Q6UXE8
Gene Alias BTN9.1; BTNL3; BTNLR; butyrophilin like 3; butyrophilin-like 3; butyrophilin-like protein 3; Butyrophilin-like receptor; COLF4100; UNQ744/PRO1472
Gene Symbols BTNL3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human BTNL3 (aa 71-120).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10917
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less