missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTEX - Clay 1, MilliporeSigma™ Supelco™
Description
Featured Industries: Environmental
Specifications
Specifications
| Chemical Name or Material | Clay |
| CAS | 14808-60-7 |
| Type | Certified Reference Material |
| Quantity | 30 g |
| Grade | Certified Reference Material |