missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, Anhydrous, 97%, Spectrum™ Chemical

Catalog No. 18613954
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613954 500 g Poly Pail
18613952 2.5 kg Poly Pail
18613951 12 kg Poly Pail
18613953 45 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18613954 Supplier Spectrum Chemical Mfg Cor Supplier No. S1498500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7772-98-7

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 500 g
Molecular Weight (g/mol) 158.11
Percent Purity 97%
Grade Reagent
Packaging Poly Pail
Chemical Name or Material Sodium Thiosulfate, Anhydrous

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.