missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BST-2 Monoclonal Antibody (K29052_18D3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA556177
Description
Sequence of this protein is as follows: QQELTEAQKG FQDVEAQAAT CNHTVMALMA SLDAEKAQGQ KKVEELEGEI TTLNHKLQDA SAEVERLRRE NQVLSVRIAD KKYYPSSQDS
BST2 (CD317, HM1.24 antigen, DAMP-2, tetherin) is an integral membrane protein that is involved in the development and growth of B-cells. The surface expression of BST2 on fibroblast cell lines facilitated the stromal cell-dependent growth of DW34, a pre-B-cell line. BST2 is highly expressed during B-cell development, from pro-B precursors to plasma cells, T-cells, monocytes, NK cells and DCs (at protein level), and myeloma cells. Research studies have shown that BST2 also functions as an inhibitor of retrovirus release from human cells whose activity is antagonized by the HIV-1 accessory protein, Vpu. While BST2 causes retention of virions on cell surfaces or endocytosis into BST2-positive compartments, its depletion abolished the viral requirement for Vpu for virus release, an activity that may represent a potential therapeutic strategy for the treatment of HIV/AIDS. Another disease associated with BST2 dysfunction is stomatitis.Chemical Identifiers
| BST-2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 2310015I10Rik; bone marrow stromal antigen 2; bone marrow stromal cell antigen 2; Bst2; BST-2; C87040; CD317; Damp1; DAMP-1; DAMP-1 protein homolog; GREG; HM1.24 antigen; NPC-A-7; PDCA-1; protein DAMP-1; TETHERIN | |
| Recombinant human BST2 (immunogen range is 71-160). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Bst2 |
| K29052_18D3 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q10589 | |
| 684 | |
| IgG1 | |
| Protein A | |
| Antibody |
Specifications
| BST-2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q10589 | |
| Bst2 | |
| Recombinant human BST2 (immunogen range is 71-160). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K29052_18D3 | |
| Unconjugated | |
| Bst2 | |
| 2310015I10Rik; bone marrow stromal antigen 2; bone marrow stromal cell antigen 2; Bst2; BST-2; C87040; CD317; Damp1; DAMP-1; DAMP-1 protein homolog; GREG; HM1.24 antigen; NPC-A-7; PDCA-1; protein DAMP-1; TETHERIN | |
| Mouse | |
| Protein A | |
| RUO | |
| 684 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |