missing translation for 'onlineSavingsMsg'
Learn More

Brucine Sulfate Heptahydrate 98.0+%, TCI America™

Catalog No. B067225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B067225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B067225G Supplier TCI America Supplier No. B067225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60583-39-3
Molecular Formula C46H68N4O19S
Molecular Weight (g/mol) 1013.12
MDL Number MFCD00150159
InChI Key PPJIVFYMRNHHTJ-UMRIXEKQSA-N
Synonym brucinesulfateheptahydrate fornitrateanalysis
PubChem CID 126969999
IUPAC Name bis((11S,18S,20R,21R,22S)-4,5-dimethoxy-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2(7),3,5,14-tetraen-9-one) sulfuric acid heptahydrate
SMILES O.O.O.O.O.O.O.OS(O)(=O)=O.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34

Specifications

Melting Point 180°C
Color White-Yellow
Quantity 25 g
UN Number 1544
Formula Weight 887.01
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Brucine Sulfate Heptahydrate [for Nitrate Analysis]
RTECSNumber WL2103350
TSCA No