accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OH2OH2OH2OCH3ON(S)(R)(S)OON(S)(R)OCH3CH3ON(S)(R)(S)OON(S)(R)OCH3OHSOHOO

CAS RN 60583-39-3

IUPAC Name: bis((11S,18S,20R,21R,22S)-4,5-dimethoxy-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2(7),3,5,14-tetraen-9-one) sulfuric acid heptahydrate
Synonyms: bis((11S,18S,20R,21R,22S)-4,5-dimethoxy-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2(7),3,5,14-tetraen-9-one) sulfuric acid heptahydrate
Molecular Weight (g/mol): 1013.12
Molecular Formula: C46H68N4O19S
InChi Key: PPJIVFYMRNHHTJ-UMRIXEKQSA-N
SMILES: O.O.O.O.O.O.O.OS(O)(=O)=O.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (3)
Grade 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)