missing translation for 'onlineSavingsMsg'
Learn More

Brucine sulfate heptahydrate, 98+%, ACS reagent

Catalog No. AC403290100
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC403290100 10 g Glass bottle
AC403290250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC403290100 Supplier Thermo Scientific Chemicals Supplier No. 403290100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 60583-39-3 | C46H68N4O19S | 1013.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60583-39-3
Molecular Formula C46H68N4O19S
Molecular Weight (g/mol) 1013.12
MDL Number MFCD00150159
InChI Key PPJIVFYMRNHHTJ-UMRIXEKQSA-N
Synonym brucinesulfateheptahydrate fornitrateanalysis
PubChem CID 126969999
IUPAC Name (4aR,5aR,8aR,13aR,15aS,15bR)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;sulfuric acid;heptahydrate
SMILES O.O.O.O.O.O.O.OS(O)(=O)=O.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34

Specifications

Color White
Ignition Residue 0.1% max.
Loss on Drying 13.0% max. (1 g, 105°C, 6 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 10 g
Sensitivity (to nitrate) Passes Test
Merck Index 15, 1464
Formula Weight 1013.13
Percent Purity 98+%
Clarity Passes test
Grade ACS Reagent
Physical Form Crystalline Powder
Chemical Name or Material Brucine sulfate heptahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal if swallowed.
Harmful to aquatic life with long lasting effects.
Fatal if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-432-9
TSCA TSCA

RUO – Research Use Only