missing translation for 'onlineSavingsMsg'
Learn More

Brucine dihydrate, 99%

Catalog No. p-77742
Change view
Click to view available options
Quantity:
50 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC163500500 50 g Glass bottle
AC163501000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC163500500 Supplier Thermo Scientific Chemicals Supplier No. 163500500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 145428-94-0 | C23H27N2O4 | 395.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 145428-94-0
Molecular Formula C23H27N2O4
Molecular Weight (g/mol) 395.48
MDL Number MFCD00005942
InChI Key RRKTZKIUPZVBMF-IBTVXLQLSA-O
Synonym brucine dihydrate, 10,11-dimethoxystrychnine, 10,11-dimethoxystrychnine dihydrate
PubChem CID 69897601
IUPAC Name (4aR,5aS,8aR,13aS,15aS,15bR)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;dihydrate
SMILES COC1=CC2=C(C=C1OC)[C@@]13CC[NH+]4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34

Specifications

Melting Point 174°C to 179°C
Color White to Beige
Loss on Drying 6.0% to 9.5% (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 50 g
Merck Index 14, 1455
Specific Rotation -123
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,chloroform and benzene,slightly soluble in ether,glycerol and,ethyl acetate
Formula Weight 430.5
Percent Purity 99%
Physical Form Crystals or Powder
Chemical Name or Material Brucine dihydrate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful to aquatic life with long lasting effects.
Fatal if swallowed.
Fatal if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only