Learn More
Thermo Scientific Chemicals Brooker's merocyanine dye
CAS: 23302-83-2 | C14H13NO | 211.26 g/mol
Supplier: Thermo Scientific Chemicals H6651703
Description
Brooker's merocyanine dye is used as solvent polarity indicators. It is used as pH sensors and transition metal cation indicators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBrooker′s merocyanine dye is used as solvent polarity indicators. It is used as pH sensors and transition metal cation indicators.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications
| Brooker's merocyanine dye | |
| 220°C (decomposition) | |
| MFCD00010099 | |
| brooker's merocyanine, 4-2-1-methylpyridin-4 1h-ylidene ethylidene cyclohexa-2,5-dien-1-one, 4-2-1-methylpyridin-4-ylidene ethylidene cyclohexa-2,5-dien-1-one, 2,5-cyclohexadien-1-one,4-2-1-methyl-4 1h-pyridinylidene ethylidene, brooker/'s merocyanine, 1-methyl-4-4-oxylatostyryl pyridinium, 1-methyl-4-e-4-oxylatostyryl pyridinium, 4-2-1-methylpyridin-4-ylidene ethylidene cyclohexa-2,5-die, 4-2-1-methyl-4-pyridylidene ethylidene cyclohexa-2,5-dien-1-one | |
| CN1C=CC(=CC=C2C=CC(=O)C=C2)C=C1 | |
| 211.26 | |
| 211.26 |
| 23302-83-2 | |
| C14H13NO | |
| 1 g | |
| DBOHWMPKJCJANT-UHFFFAOYSA-N | |
| 4-[2-(1-methylpyridin-4-ylidene)ethylidene]cyclohexa-2,5-dien-1-one | |
| 258436 |
Chemical Identifiers
| 23302-83-2 | |
| 211.26 | |
| DBOHWMPKJCJANT-UHFFFAOYSA-N | |
| 258436 | |
| CN1C=CC(=CC=C2C=CC(=O)C=C2)C=C1 |
| C14H13NO | |
| MFCD00010099 | |
| brooker's merocyanine, 4-2-1-methylpyridin-4 1h-ylidene ethylidene cyclohexa-2,5-dien-1-one, 4-2-1-methylpyridin-4-ylidene ethylidene cyclohexa-2,5-dien-1-one, 2,5-cyclohexadien-1-one,4-2-1-methyl-4 1h-pyridinylidene ethylidene, brooker/'s merocyanine, 1-methyl-4-4-oxylatostyryl pyridinium, 1-methyl-4-e-4-oxylatostyryl pyridinium, 4-2-1-methylpyridin-4-ylidene ethylidene cyclohexa-2,5-die, 4-2-1-methyl-4-pyridylidene ethylidene cyclohexa-2,5-dien-1-one | |
| 4-[2-(1-methylpyridin-4-ylidene)ethylidene]cyclohexa-2,5-dien-1-one |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only