accessibility menu, dialog, popup

UserName

CAS RN 23302-83-2

IUPAC Name: 4-[2-(1-methyl-1,4-dihydropyridin-4-ylidene)ethylidene]cyclohexa-2,5-dien-1-one
Synonyms: brooker's merocyanine
Molecular Weight (g/mol): 211.26
Molecular Formula: C14H13NO
InChi Key: DBOHWMPKJCJANT-UHFFFAOYSA-N
SMILES: CN1C=CC(=CC=C2C=CC(=O)C=C2)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)