missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brooker's merocyanine dye

Catalog No. AAH66517MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66517MD 250 mg
AAH6651703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66517MD Supplier Thermo Scientific Chemicals Supplier No. H66517MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 23302-83-2 | C14H13NO | 211.26 g/mol

Brooker's merocyanine dye is used as solvent polarity indicators. It is used as pH sensors and transition metal cation indicators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brooker′s merocyanine dye is used as solvent polarity indicators. It is used as pH sensors and transition metal cation indicators.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Chemical Identifiers

CAS 23302-83-2
Molecular Formula C14H13NO
Molecular Weight (g/mol) 211.26
MDL Number MFCD00010099
InChI Key DBOHWMPKJCJANT-UHFFFAOYSA-N
Synonym brooker's merocyanine, 4-2-1-methylpyridin-4 1h-ylidene ethylidene cyclohexa-2,5-dien-1-one, 4-2-1-methylpyridin-4-ylidene ethylidene cyclohexa-2,5-dien-1-one, 2,5-cyclohexadien-1-one,4-2-1-methyl-4 1h-pyridinylidene ethylidene, brooker/'s merocyanine, 1-methyl-4-4-oxylatostyryl pyridinium, 1-methyl-4-e-4-oxylatostyryl pyridinium, 4-2-1-methylpyridin-4-ylidene ethylidene cyclohexa-2,5-die, 4-2-1-methyl-4-pyridylidene ethylidene cyclohexa-2,5-dien-1-one
PubChem CID 258436
IUPAC Name 4-[2-(1-methylpyridin-4-ylidene)ethylidene]cyclohexa-2,5-dien-1-one
SMILES CN1C=CC(=CC=C2C=CC(=O)C=C2)C=C1

Specifications

Melting Point 220°C (decomposition)
Quantity 250 mg
Formula Weight 211.26
Chemical Name or Material Brooker's merocyanine dye
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only