missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromotrimethylsilane 95.0+%, TCI America™
Supplier: TCI America B10875ML
Specifications
| Bromotrimethylsilane | |
| -43°C | |
| 79°C | |
| MFCD00000048 | |
| 2920 | |
| IYYIVELXUANFED-UHFFFAOYSA-N | |
| bromo(trimethyl)silane | |
| 76113 | |
| ≥95.0% (GC) |
| 2857-97-8 | |
| Yellow | |
| C3H9BrSi | |
| 5 mL | |
| trimethylsilyl bromide, trimethylbromosilane, silane, bromotrimethyl, trimethylsilicon bromide, tmsbr, trimethylsilylbromide, bromotrimethyl silane, tmbs, bromo trimethyl silane, tms bromide | |
| C[Si](C)(C)Br | |
| 153.094 | |
| 153.09 | |
| Liquid |
Chemical Identifiers
| 2857-97-8 | |
| 153.094 | |
| IYYIVELXUANFED-UHFFFAOYSA-N | |
| 76113 | |
| C[Si](C)(C)Br |
| C3H9BrSi | |
| MFCD00000048 | |
| trimethylsilyl bromide, trimethylbromosilane, silane, bromotrimethyl, trimethylsilicon bromide, tmsbr, trimethylsilylbromide, bromotrimethyl silane, tmbs, bromo trimethyl silane, tms bromide | |
| bromo(trimethyl)silane |
Safety and Handling
TSCA : Yes