missing translation for 'onlineSavingsMsg'
Learn More

Bromotrimethylsilane 95.0+%, TCI America™

Catalog No. p-7118843
Change view
Click to view available options
Quantity:
5 mL
25 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10875ML 5 mL
B108725ML 25 mL
B1087250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B10875ML Supplier TCI America Supplier No. B10875ML
Only null left

Chemical Identifiers

CAS 2857-97-8
Molecular Formula C3H9BrSi
Molecular Weight (g/mol) 153.094
MDL Number MFCD00000048
InChI Key IYYIVELXUANFED-UHFFFAOYSA-N
Synonym trimethylsilyl bromide, trimethylbromosilane, silane, bromotrimethyl, trimethylsilicon bromide, tmsbr, trimethylsilylbromide, bromotrimethyl silane, tmbs, bromo trimethyl silane, tms bromide
PubChem CID 76113
IUPAC Name bromo(trimethyl)silane
SMILES C[Si](C)(C)Br

Specifications

Melting Point -43°C
Color Yellow
Boiling Point 79°C
Quantity 5 mL
UN Number 2920
Formula Weight 153.09
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromotrimethylsilane