missing translation for 'onlineSavingsMsg'
Learn More

Bromotrichloromethane 98.0+%, TCI America™

Catalog No. B066225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B066225G 25 g
B0662500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B066225G Supplier TCI America Supplier No. B066225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-62-7
Molecular Formula CBrCl3
Molecular Weight (g/mol) 198.265
MDL Number MFCD00000783
InChI Key XNNQFQFUQLJSQT-UHFFFAOYSA-N
Synonym methane, bromotrichloro, trichlorobromomethane, bromo trichloro methane, carbon bromotrichloride, monobromotrichloromethane, carbon trichlorobromide, trichloromethyl bromide, caswell no. 118, unii-ikj30qxm63, ccl3br
PubChem CID 6383
IUPAC Name bromo(trichloro)methane
SMILES C(Cl)(Cl)(Cl)Br

Specifications

Melting Point -6°C
Color Colorless
Boiling Point 104°C
Quantity 25 g
UN Number 2810
Formula Weight 198.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromotrichloromethane
EINECSNumber (2)-2363
RTECSNumber PA5400000
TSCA Yes