accessibility menu, dialog, popup

UserName

ClClClBr

CAS RN 75-62-7

IUPAC Name: bromotrichloromethane
Synonyms: bromotrichloromethane
Molecular Weight (g/mol): 198.27
Molecular Formula: CBrCl3
InChi Key: XNNQFQFUQLJSQT-UHFFFAOYSA-N
SMILES: ClC(Cl)(Cl)Br
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)