missing translation for 'onlineSavingsMsg'
Learn More

Bromotrichloromethane, 99%

Catalog No. AC107491000
Change view
Click to view available options
Quantity:
100 mL
250 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107491000 100 mL Glass bottle
AC107492500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC107491000 Supplier Thermo Scientific Chemicals Supplier No. 107491000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 75-62-7 | CBrCl3 | 198.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 75-62-7
Molecular Formula CBrCl3
Molecular Weight (g/mol) 198.27
MDL Number MFCD00000783
InChI Key XNNQFQFUQLJSQT-UHFFFAOYSA-N
Synonym methane, bromotrichloro, trichlorobromomethane, bromo trichloro methane, carbon bromotrichloride, monobromotrichloromethane, carbon trichlorobromide, trichloromethyl bromide, caswell no. 118, unii-ikj30qxm63, ccl3br
PubChem CID 6383
IUPAC Name bromo(trichloro)methane
SMILES C(Cl)(Cl)(Cl)Br

Specifications

Melting Point -6.0°C
Color Colorless to Yellow
Density 1.9970g/mL
Boiling Point 105.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5055 to 1.5075
Linear Formula BrCCl3
Quantity 100 mL
Beilstein 01, 67
Fieser 01,80; 02,42; 07,40
Specific Gravity 1.997
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with many organic liquids, soluble in ethanol and ether
Formula Weight 198.27
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Bromotrichloromethane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
EINECSNumber 200-886-0
RTECSNumber PA5400000
TSCA TSCA

RUO – Research Use Only