missing translation for 'onlineSavingsMsg'
Learn More

Bromotrichloromethane, 97%

Catalog No. p-7081031
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1082422 100 g
AAA1082436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1082422 Supplier Thermo Scientific Chemicals Supplier No. A1082422

missing translation for 'validMainframeMsg'

CAS: 75-62-7 | CBrCl3 | 198.265 g/mol

Used in the bromination of benzylic positions under sunlamp irradiation. Employed as chain transfer agent for radical polymerization of methacrylates. It can also be employed as brominating reagent of active hydrogen compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the bromination of benzylic positions under sunlamp irradiation. Employed as chain transfer agent for radical polymerization of methacrylates. It can also be employed as brominating reagent of active hydrogen compounds.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Protect against electrostatic charges.

Chemical Identifiers

CAS 75-62-7
Molecular Formula CBrCl3
Molecular Weight (g/mol) 198.265
MDL Number MFCD00000783
InChI Key XNNQFQFUQLJSQT-UHFFFAOYSA-N
Synonym methane, bromotrichloro, trichlorobromomethane, bromo trichloro methane, carbon bromotrichloride, monobromotrichloromethane, carbon trichlorobromide, trichloromethyl bromide, caswell no. 118, unii-ikj30qxm63, ccl3br
PubChem CID 6383
IUPAC Name bromo(trichloro)methane
SMILES C(Cl)(Cl)(Cl)Br

Specifications

Melting Point -6°C
Density 2.015
Boiling Point 104°C to 105°C
Flash Point None
Odor Chloroform-like
Refractive Index 1.506
Linear Formula BrCCl3
Quantity 100 g
UN Number UN2810
Beilstein 1732543
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 198.28
Percent Purity 97%
Chemical Name or Material Bromotrichloromethane
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 200-886-0
RTECSNumber PA5400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only