missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue, Water Soluble, ACS, Spectrum™ Chemical

$254.69 - $6,387.56

Chemical Identifiers

CAS 34722-90-2
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18609272
View Documents Promotion Details
N/A 5 g Amber Glass Bottle
Each for $254.69
Only null left
 
18609273
View Documents Promotion Details
N/A 25 g Amber Glass Bottle
Each for $671.47
Only null left
 
18609274
View Documents Promotion Details
N/A 100 g Amber Glass Bottle
Each for $2,483.16
Only null left
 
18609275
View Documents Promotion Details
N/A 1 kg Amber Glass Bottle
Each for $6,387.56
Only null left
 
Description

Description

Specifications

Chemical Identifiers

34722-90-2

Specifications

34722-90-2
ACS
Bromothymol Blue
1
Orange/Pink to Red or Purple
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.