missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue, Water Soluble, ACS, Spectrum™ Chemical

Catalog No. 18609272
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609272 5 g Amber Glass Bottle
18609273 25 g Amber Glass Bottle
18609274 100 g Amber Glass Bottle
18609275 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609272 Supplier Spectrum Chemical Mfg Cor Supplier No. BR1555GM

Chemical Identifiers

CAS 34722-90-2

Specifications

CAS Max % 1
Grade ACS
Packaging Amber Glass Bottle
Color Orange/Pink to Red or Purple
Quantity 5 g
Chemical Name or Material Bromothymol Blue

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.