missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Bromothymol Blue sodium salt
CAS: 34722-90-2 | C27H27Br2NaO5S | 646.37 g/mol
$88.08 - $255.92
Chemical Identifiers
| CAS | 34722-90-2 |
|---|---|
| Molecular Formula | C27H27Br2NaO5S |
| Molecular Weight (g/mol) | 646.37 |
| MDL Number | MFCD00077263,MFCD00077263,MFCD00077263 |
| InChI Key | NMKFVGALBGZKGW-FKWCIMQXSA-M |
| Synonym | Bromothymol Blue, water soluble |
| PubChem CID | 102183223 |
| IUPAC Name | sodium;2-bromo-4-[(Z)-(3-bromo-2-methyl-4-oxo-5-propan-2-ylcyclohexa-2,5-dien-1-ylidene)-(2-sulfophenyl)methyl]-3-methyl-6-propan-2-ylphenolate |
| SMILES | [Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAA1791309
|
Thermo Scientific Chemicals
A1791309 |
10 g |
Each for $88.08
|
|
|||||
|
AAA1791318
|
Thermo Scientific Chemicals
A1791318 |
50 g |
Each for $255.92
|
|
|||||
Description
It is employed as a pH indicator which is yellow at pH ? 6.0 and blue at pH ? 7.6. It finds its major application in lithium battery, textiles and antagonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a pH indicator which is yellow at pH ≤ 6.0 and blue at pH ≥ 7.6. It finds its major application in lithium battery, textiles and antagonists.
Solubility
Soluble in water and ethanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 34722-90-2 | |
| 646.37 | |
| NMKFVGALBGZKGW-FKWCIMQXSA-M | |
| 102183223 | |
| [Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O |
| C27H27Br2NaO5S | |
| MFCD00077263,MFCD00077263,MFCD00077263 | |
| Bromothymol Blue, water soluble | |
| sodium;2-bromo-4-[(Z)-(3-bromo-2-methyl-4-oxo-5-propan-2-ylcyclohexa-2,5-dien-1-ylidene)-(2-sulfophenyl)methyl]-3-methyl-6-propan-2-ylphenolate |
Specifications
| 200°C to 202°C | |
| C27H27Br2NaO5S | |
| Bromothymol Blue, water soluble | |
| NMKFVGALBGZKGW-FKWCIMQXSA-M | |
| sodium;2-bromo-4-[(Z)-(3-bromo-2-methyl-4-oxo-5-propan-2-ylcyclohexa-2,5-dien-1-ylidene)-(2-sulfophenyl)methyl]-3-methyl-6-propan-2-ylphenolate | |
| 102183223 | |
| Bromothymol Blue sodium salt |
| 34722-90-2 | |
| MFCD00077263,MFCD00077263,MFCD00077263 | |
| Soluble in water and ethanol. | |
| [Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O | |
| 646.37 | |
| 646.38 |
Safety and Handling
EINECSNumber : 252-169-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only