missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromothymol Blue sodium salt, 0.04% w/v aq. soln.
CAS: 34722-90-2 | C27H27Br2NaO5S | 646.37 g/mol
$50.66 - $92.27
Chemical Identifiers
| CAS | 34722-90-2 |
|---|---|
| Molecular Formula | C27H27Br2NaO5S |
| Molecular Weight (g/mol) | 646.37 |
| MDL Number | MFCD00077263,MFCD00077263,MFCD00077263 |
| InChI Key | NMKFVGALBGZKGW-FKWCIMQXSA-M |
| PubChem CID | 102183223 |
| SMILES | [Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 34722-90-2 | |
| 646.37 | |
| NMKFVGALBGZKGW-FKWCIMQXSA-M | |
| [Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O |
| C27H27Br2NaO5S | |
| MFCD00077263,MFCD00077263,MFCD00077263 | |
| 102183223 |
Specifications
| 34722-90-2 | |
| MFCD00077263,MFCD00077263,MFCD00077263 | |
| [Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O | |
| 646.37 | |
| 646.38 | |
| Blue | |
| Bromothymol Blue sodium salt |
| C27H27Br2NaO5S | |
| NMKFVGALBGZKGW-FKWCIMQXSA-M | |
| sodium;2-bromo-4-[(Z)-(3-bromo-2-methyl-4-oxo-5-propan-2-ylcyclohexa-2,5-dien-1-ylidene)-(2-sulfophenyl)methyl]-3-methyl-6-propan-2-ylphenolate | |
| 102183223 | |
| Odorless | |
| Liquid |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only